Arseen ≥99.99999% (metals basis), polycrystalline lumps, Puratronic® 2-8 mm (0.08-0.3 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


Packaged under argon.

13044.03 13044.09 13044.14 13044.22 13044.A1
13044.03EA 33.6 EUR
13044.03 13044.09 13044.14 13044.22 13044.A1
Arseen ≥99.99999% (metals basis), polycrystalline lumps, Puratronic® 2-8 mm (0.08-0.3 in)
Arseen

Purity: 99.99999+% CAS No.: 7440-38-2 Molecular Formula: As Formula Weight: 74.92 Physical Form: Packaged under argon Sensitivity: Air Sensitive Appearance: Grey Odour: Odourless UN No.: 1558 MDL No.: MFCD00085309

Formule: As
Smeltpunt: 817 °C
Dichtheid: 5,727 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00085309
CAS nummer: 7440-38-2
EINECS: 213-148-6
UN: 1558
ADR: 6.1,II
Merck Index: 12,00832

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR