VWR® Blue Line, Laboratory Chairs and Stool
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
132-1375EA 157.7 EUR
132-1375 132-1376 132-1377 132-1378 132-1379
VWR® Blue Line, Laboratory Chairs and Stool
Stoelen
These robust chairs and stool are designed for use in laboratories and are perfect for many applications from pharmaceutical, biotech, chemicals, healthcare and cleanroom to medical technology. The seat is made from artificial and antibacterial leather, and the backrest is made of plastic in such a way that there are no joints. This means that the unit is extremely easy to keep clean.
  • 3-D flex function on the front edge of the seat, and the backrest automatically follows
  • Solid plastic star base in black
  • Coloured closed seat shell
  • Easy to clean, washable
More Product Information
Limited Time Sale
Select items below are currently on sale