Kaliumpermanganaat 99-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

13206-1KG 13206-25KG 13206-2.5KG 13206-250G
13206-1KGEA 113 EUR
13206-1KG 13206-25KG 13206-2.5KG 13206-250G
Kaliumpermanganaat 99-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
Kaliumpermanganaat
Formule: KMnO₄
Smeltpunt: Dec.
Dichtheid: 2,700 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011364
CAS nummer: 7722-64-7
EINECS: 231-760-3
UN: 1490
ADR: 5.1,II
Merck Index: 12,07824

Order Now

Specification Test Results

Assay 99.0 - 100.5 %
Assay (calculated to the dried substance) 99.0 - 100.5 %
Water insoluble matter Max. 0.2 %
Loss on drying (on silica gel, 18h) Max. 0.2 %
Chloride (Cl) Max. 100 ppm
Sulfate (SO₄) Max. 500 ppm
Appearance of the solution complying
Residual solvents complying

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR