Zinc selenide ≥99.995% (metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

13241.18 13241.30
13241.18EA 123 EUR
13241.18 13241.30
Zinc selenide ≥99.995% (metals basis)
Zinc selenide

Purity: 99.995% CAS No.: 1315-09-9 Molecular Formula: ZnSe Formula Weight: 144.33 Physical Form: 3-12mm Lump Density: 5.42 Melting Point: 1100° Merck Reference: 14,10156 UN No.: 3283 MDL No.: MFCD00011296

Formule: SeZn
Smeltpunt: >1100 °C
Dichtheid: 5,42
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011296
CAS nummer: 1315-09-9
EINECS: 215-259-7
UN: 3283
ADR: 6.1,III
Merck Index: 14,10156

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR