Platinum rhodium alloy (90:10 w%), wire for ISA type S standard grade thermocouple, Ø 0.5 mm (0.02 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

13372.BU 13372.BY
13372.BUEA 667 EUR
13372.BU 13372.BY
Platinum rhodium alloy (90:10 w%), wire for ISA type S standard grade thermocouple, Ø 0.5 mm (0.02 in)
Platinum rhodium alloy (90:10 w%)

Molecular Formula: Pt:Rh; 90:10 wt% Physical Form: ≈1.04g/25cm Odour: Odourless MDL No.: MFCD00198186

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR