Digallium trisulphide ≥99.99% (metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

13387.04 13387.09 13387.02
13387.04EA 145 EUR
13387.04 13387.09 13387.02
Digallium trisulphide ≥99.99% (metals basis)
Digallium trisulphide

Purity: 99.99% CAS No.: 12024-22-5 Molecular Formula: Ga2S3, may contain small amount of oxide Formula Weight: 235.63 Physical Form: Powder Density: 3.77 Melting Point: 1255° UN No.: 3260 MDL No.: MFCD00016106

Formule: Ga₂S₃
Smeltpunt: 1255 °C
Dichtheid: 3,77
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00016106
CAS nummer: 12024-22-5
EINECS: 234-692-2
UN: 3260
ADR: 8,II

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR