Disodium hydrogen phosphate, watervrij ≥99.0% ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

13437.30 13437.A1 13437.A7
13437.30EA 25.3 EUR
13437.30 13437.A1 13437.A7
Disodium hydrogen phosphate, watervrij ≥99.0% ACS
Disodium hydrogen phosphate

Purity: 99.0% min CAS No.: 7558-79-4 Molecular Formula: HNa2O4P Formula Weight: 141.98 Physical Form: Granular Sensitivity: Hygroscopic Merck Reference: 14,8659 MDL No.: MFCD00003496

Formule: Na₂HPO₄
Smeltpunt: 250 °C
Dichtheid: 1,53 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003496
CAS nummer: 7558-79-4
EINECS: 231-448-7
Bereik: 01-2119489797-11
Merck Index: 14,8659

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR