Sodium thiosulphate pentahydrate 99-101%, Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

13479-5KG 13479-500G 13479-2.5KG 13479-4X2.5KG
13479-5KGEA 181 EUR
13479-5KG 13479-500G 13479-2.5KG HONC13479-4X2.5KG
Sodium thiosulphate pentahydrate 99-101%, Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
Sodium thiosulphate pentahydrate
Formule: Na₂S₂O₃.5H₂O
Kookpunt: Decomposes on heating,
Dichtheid: 1,73 g/cm³ (20 °C)
Bewaar temperatuur: Room Temperature
MDL Nummer: MFCD00149186
CAS nummer: 10102-17-7
EINECS: 231-867-5
Merck Index: 12,08844

Order Now

Specification Test Results

Identity complying
Assay 99.0 - 101.0 %
Assay (calculated on the anhydrous basis) 99.0 - 100.5 %
Loss on drying (vacuum, 16 h) 32.0 - 37.0 %
pH (10 %, 20°C) 6.0 - 9.0
Arsenic (As) Max. 0.0003 %
Calcium (Ca) Max. 0.005 %
Iron (Fe) Max. 0.001 %
Heavy metals (as Pb) Max. 0.0005 %
Chloride (Cl) Max. 0.05 %
Sulfate and sulfite (as SO₄) Max. 0.2 %
Sulfide (S) complying
Appearance of the solution complying
Residual solvents complying

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR