Rubidium iodide ≥99.8% (metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

13497.09 13497.18 13497.A7
13497.09EA 79.9 EUR
13497.09 13497.18 13497.A7
Rubidium iodide ≥99.8% (metals basis)
Rubidium iodide

Purity: 99.8% CAS No.: 7790-29-6 Molecular Formula: RbI Formula Weight: 212.37 Physical Form: Crystalline Density: 3.55 Refractive Index: 1.6474 Sensitivity: Hygroscopic Odour: Odourless Melting Point: 647° Boiling Point: 1300° Solubility: Freely soluble in water. Soluble in alcohol Merck Reference: 14,8286 MDL No.: MFCD00011192

Formule: IRb
Kookpunt: 1300 °C
Smeltpunt: 647 °C
Dichtheid: 3,55
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011192
CAS nummer: 7790-29-6
EINECS: 232-198-1
Merck Index: 14,8286

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR