Pharmaceutical exhausts
Leverancier: asecos
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
31833 31834
135-1772EA 2720 EUR
135-1772 135-1773
Pharmaceutical exhausts
Zuurkasten en behuizingen Farmaceutische afzuigkappen
For safe handling of chemicals, protection from hazardous vapours and potentially explosive atmospheres. Hazardous material workplace (pharmaceutical exhausts).
  • Easily mounted on a table top
  • Robust construction made of chemically resistant, antibacterial plastic laminated sheet steel
  • Exhaust air monitoring with visual and audible alarm
Standard equipment: exhaust air monitoring, 2 sockets, workplace lighting, acrylic glass sliding windows with continuously adjustable height, exhaust air fan. Accessories like media supplies, support frame andMore Product Information