Safety storage cabinets, type 90, V-MOVE-90
Leverancier: asecos
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
35034001 35034005 35035001 35035005 35085040 35085041 35085051 35085047 35085046 35086040 35086041 35086051 35086047 35086046
135-2636EA 4690 EUR
135-2636 135-2637 135-2638 135-2639 135-2640 135-2641 135-2644 135-2643 135-2642 135-2645 135-2646 135-2649 135-2648 135-2647
Safety storage cabinets, type 90, V-MOVE-90
Meubilair Opslagkasten
Using the vertical drawer technology, the V-MOVE-90 safety storage cabinets have the same storage volume as standard cabinets with only half the corpus width.
  • Robust construction and longevity: robust, high-quality, scratch-resistant outer body with powder-coated surface
  • Electronic opening and closing of drawers at the push of a button
  • Vertical drawer provides an overview and simple access to the stored containers
  • Serial automatic door closing (TSA) after approx. 60 seconds with acoustic and visual signal before drawer closing
  • No unauthorised use: vertical
More Product Information