Erbium trifluoride, anhydrous ≥99.9% (REO, rare earth oxide basis), REacton®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

13653.09 13653.18
13653.09EA 102 EUR
13653.09 13653.18
Erbium trifluoride, anhydrous ≥99.9% (REO, rare earth oxide basis), REacton®
Erbium trifluoride

Purity: 99.9% CAS No.: 13760-83-3 Molecular Formula: ErF3 Formula Weight: 224.28 Physical Form: Powder Density: 6.4 Melting Point: 1350° MDL No.: MFCD00016074

Formule: ErF₃
Kookpunt: 2200 °C (1013 hPa)
Smeltpunt: 1350 °C
Dichtheid: 6,4
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00016074
CAS nummer: 13760-83-3
EINECS: 237-356-3
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR