Bariumchloride, ultra dry ≥99.998% (metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


Ampuled under argon

13686.14
13686.14EA 193 EUR
13686.14
Bariumchloride, ultra dry ≥99.998% (metals basis)
Bariumchloride

Purity: 99.998% CAS No.: 10361-37-2 Molecular Formula: BaCl2 Formula Weight: 208.25 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 3.9 Appearance: White Odour: Odourless Melting Point: 963° Boiling Point: 1560° Merck Reference: 14,971 UN No.: 1564 MDL No.: MFCD00003445

Formule: BaCl₂
MW: 208.24 g/mol
Kookpunt: 1560 °C
Smeltpunt: 963 °C
Dichtheid: 3.9
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003445
CAS nummer: 10361-37-2
EINECS: 233-788-1
UN: 1564
ADR: 6.1 III
Merck Index: 14,971

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR