Rhenium standard solution, 1000 mg/L Re in 5% nitric acid (from Re), standard for ICP, Specpure®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

13817.AD 13817.AE 13817.AP
13817.ADEA 104 EUR
13817.AD 13817.AE 13817.AP
Rhenium standard solution, 1000 mg/L Re in 5% nitric acid (from Re), standard for ICP, Specpure®
Standaarden Kationische standaarden Renium standaarden

Molecular Formula: Re in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011195


Certificaten: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR