Bismuth standard solution, 1000 mg/L Bi in 5% nitric acid (from Bi), standard for ICP, Specpure®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

13822.AD 13822.AE 13822.AP
13822.ADEA 52.4 EUR
13822.AD 13822.AE 13822.AP
Bismuth standard solution, 1000 mg/L Bi in 5% nitric acid (from Bi), standard for ICP, Specpure®
Standaarden Kationische standaarden Bismut standaarden

Molecular Formula: Bi in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134033


Certificaten: Traceable to NIST

Order Now

Specificaties

Environmentally Preferable
MDL No. MFCD00134033

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR