Thulium standard solution, 1000 mg/L Tm in 5% nitric acid (from Tm₂O₃), standard for ICP, Specpure®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

13824.AD 13824.AE 13824.AP
13824.ADEA 87.5 EUR
13824.AD 13824.AE 13824.AP
Thulium standard solution, 1000 mg/L Tm in 5% nitric acid (from Tm₂O₃), standard for ICP, Specpure®
Standaarden Kationische standaarden Thulium standaarden

Molecular Formula: Tm₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011281


Certificaten: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR