Platinum standard solution, 1000 mg/L Pt in 20% hydrochloric acid (from Pt), standard for ICP, Specpure®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

13827.AD 13827.AE 13827.AP
13827.ADEA 105 EUR
13827.AD 13827.AE 13827.AP
Platinum standard solution, 1000 mg/L Pt in 20% hydrochloric acid (from Pt), standard for ICP, Specpure®
Standaarden Kationische standaarden Platina standaarden

Molecular Formula: Pt in 20% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00011179


Certificaten: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR