Palladium standard solution, 1000 mg/L Pd in 20% hydrochloric acid (from Pd), standard for ICP, Specpure®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

13833.AD 13833.AE 13833.AP
13833.ADEA 114 EUR
13833.AD 13833.AE 13833.AP
Palladium standard solution, 1000 mg/L Pd in 20% hydrochloric acid (from Pd), standard for ICP, Specpure®
Standaarden Kationische standaarden Palladium standaarden

Molecular Formula: Pd in 20% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD03613603


Certificaten: Traceable to NIST

Order Now

Specificaties

Environmentally Preferable
MDL No. MFCD03613603

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR