Dysprosium standard solution, 1000 mg/L Dy in 5% nitric acid (from Dy₂O₃), standard for ICP, Specpure®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

13838.AD 13838.AE 13838.AP
13838.ADEA 76.3 EUR
13838.AD 13838.AE 13838.AP
Dysprosium standard solution, 1000 mg/L Dy in 5% nitric acid (from Dy₂O₃), standard for ICP, Specpure®
Standaarden Kationische standaarden Dysprosium standaarden

Molecular Formula: Dy₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010982


Certificaten: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR