Beryllium standard solution, 1000 mg/L Be in 5% nitric acid (from BeO.(Be(OAc)₂)₃), standard for ICP, Specpure®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

13848.ADEA 51.8 EUR
13848.AD 13848.AE
Beryllium standard solution, 1000 mg/L Be in 5% nitric acid (from BeO.(Be(OAc)₂)₃), standard for ICP, Specpure®
Standaarden Kationische standaarden Beryllium standaarden

Molecular Formula: Be₄O(C₂H₃O₂)6 in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134032


Certificaten: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR