Yttrium standard solution, 1000 mg/L Y in 5% nitric acid (from Y₂O₃), standard for ICP, Specpure®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

13855.AP 13855.AE 13855.AD
13855.APEA 200 EUR
13855.AP 13855.AE 13855.AD
Yttrium standard solution, 1000 mg/L Y in 5% nitric acid (from Y₂O₃), standard for ICP, Specpure®
Standaarden Kationische standaarden Yttrium standaarden

Molecular Formula: Y₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011468


Certificaten: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR