Gallium standard solution, 1000 mg/L Ga in 5% nitric acid with hydrochloric acid (max. 1%) (from Ga), standard for ICP, Specpure®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

13869.AE 13869.AD 13869.AP
13869.AEEA 90.7 EUR
13869.AE 13869.AD 13869.AP
Gallium standard solution, 1000 mg/L Ga in 5% nitric acid with hydrochloric acid (max. 1%) (from Ga), standard for ICP, Specpure®
Standaarden Kationische standaarden Gallium standaarden

Molecular Formula: Ga in 5% HNO₃/tr. HCl
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134045


Certificaten: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR