Lanthanum standard solution, 1000 mg/L La in 5% nitric acid (from La₂O₃), standard for ICP, Specpure®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

13870.AD 13870.AE 13870.AP
13870.ADEA 51.1 EUR
13870.AD 13870.AE 13870.AP
Lanthanum standard solution, 1000 mg/L La in 5% nitric acid (from La₂O₃), standard for ICP, Specpure®
Standaarden Kationische standaarden Lanthanium standaarden

Molecular Formula: La₂O₃ in 5% HNO₃ Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011066


Certificaten: Traceable to NIST

Order Now

Specificaties

Environmentally Preferable
MDL No. MFCD00011066

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR