Titanium(IV)oxide 99-100.5%, Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

14027-1KG 14027-25KG 14027-250G
14027-1KGEA 101 EUR
14027-1KG 14027-25KG 14027-250G
Titanium(IV)oxide 99-100.5%, Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
Titanium(IV)oxide
Formule: TiO₂
Kookpunt: >2,000 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011269
CAS nummer: 13463-67-7
EINECS: 236-675-5
Merck Index: 13,09549

Order Now

Specification Test Results

Assay 98.0 - 100.5 %
Assay (calculated to the dried substance) 99.0 - 100.5 %
Water soluble matter Max. 0.25 %
Water soluble matter (hot) Max. 0.5 %
In acid (HCl 0,5 mol/l) Solution matter Max. 0.5 %
Loss on drying (105°C, 3h) Max. 0.5 %
Loss on ignition (800 °C) Max. 0.5 %
Acidity or alkalinity complying
Arsenic (As) Max. 20 ppm
Barium (Ba) complying
Iron (Fe) Max. 50 ppm
Antimony (Sb) Max. 50 ppm
Heavy metals (as Pb) Max. 20 ppm
Appearance of the solution complying
Residual solvents complying

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR