Mop, AlphaMop™, PE
Leverancier: Texwipe
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
TX7108 TX7118 TX7026 STX7118 TX7118M TX7118F TX7108H TX7123 STX7118M
129-0180EA 149 EUR
129-0180 129-0181 141-0249 141-1048 141-1060 141-1059 141-1057 141-1064 141-1049
Mop, AlphaMop™, PE
Moppen
Ideal for cleaning ceilings, walls, floors, and other flat surfaces in critical environments. The changeable, laundered polyester head fits over a flat foam swivel pad assembly attached to a fibreglass handle. Replaceable foam pad ensures the polyester head conforms to textured floors and other surfaces while resisting corrosion and abrasion. A lightweight, durable fibreglass handle gives the user excellent control over the swivel head.
  • Lightweight mop maintains ultra-low generation of particles and extractables
  • Mop can be sterilised
  • Mophead covers can
More Product Information