Acid cleaners, TICKOPUR TR 2
Leverancier: BANDELIN-ELECTRONIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
891 893
142-1906EA 23.3 EUR
142-1906 142-1985
Acid cleaners, TICKOPUR TR 2
Reinigingsmiddelen Detergenten en oppervlaktereinigers
TICKOPUR R 2 is a special cleaner based on phosphoric acid for cleaning of disassembled and non-disassembled machines, carburettors, tools, equipment and parts.
  • Gentle cleaning
  • De-emulsifying concentrate
  • Corrorsion protection
  • Weakly acid, pH 3,6
  • Dosage 0,1 - 5%
These can be used to clean metals also light metals, glass, ceramics, plastic and rubber.
Ideal for mineral residues, drifting rust, pigments, grinding-, polishing- and lapping-residues, oil, grease and wax.