Aluminum silicate

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

14231.36 14231.A3
14231.36EA 48.8 EUR
14231.36 14231.A3
Aluminum silicate
Aluminum silicate

Aluminum silicate is used to prepare aluminum-silicate polymer composite (PASiC), which is an inorganic coagulant used in water treatment. It is used as a catalyst for the pyrolysis of rice husk to crude bio -oil, which upgraded to supercritical ethanol. Further, it is used as filler in paper, rubber and in paints.


CAS No.: 12428-46-5 Molecular Formula: Al2Si2O7.2H2O Formula Weight: 258.16 (222.13anhy) Physical Form: Powder Appearance: White to pale grey Odour: Odourless MDL No.: MFCD00062311

Formule: Al₂Si₂O₇·2H₂O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00062311
CAS nummer: 12428-46-5

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR