Silver standard solution, 10000 mg/L Ag in 5% nitric acid (from Ag), standard for ICP, Specpure®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

14418.AE 14418.AD 14418.AP
14418.AEEA 164 EUR
14418.AE 14418.AD 14418.AP
Silver standard solution, 10000 mg/L Ag in 5% nitric acid (from Ag), standard for ICP, Specpure®
Standaarden Kationische standaarden Zilver standaarden

Molecular Formula: Ag in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00003397


Certificaten: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR