Barium standard solution, 10000 mg/L Ba in 5% nitric acid (from Ba(NO₃)₂), standard for ICP, Specpure®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

14423.AE 14423.AD 14423.AP
14423.AEEA 131 EUR
14423.AE 14423.AD 14423.AP
Barium standard solution, 10000 mg/L Ba in 5% nitric acid (from Ba(NO₃)₂), standard for ICP, Specpure®
Standaarden Kationische standaarden Barium standaarden

Molecular Formula: Ba(NO₃)₂ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134031


Certificaten: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR