Dysprosium standard solution, 10000 mg/L Dy in 5% nitric acid (from Dy₂O₃), standard for ICP, Specpure®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

14426.AD 14426.AE 14426.AP
14426.ADEA 102 EUR
14426.AD 14426.AE 14426.AP
Dysprosium standard solution, 10000 mg/L Dy in 5% nitric acid (from Dy₂O₃), standard for ICP, Specpure®
Standaarden Kationische standaarden Dysprosium standaarden

Molecular Formula: Dy₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010982


Certificaten: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR