Ytterbium standard solution, 10000 mg/L Yb in 5% nitric acid (from Yb₂O₃), standard for ICP, Specpure®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

14439.AE 14439.AP
14439.AEEA 155 EUR
14439.AE 14439.AP
Ytterbium standard solution, 10000 mg/L Yb in 5% nitric acid (from Yb₂O₃), standard for ICP, Specpure®
Standaarden Kationische standaarden Ytterbium standaarden

Molecular formula: Yb₂O₃ in 5% HNO₃
Physical form: liquid
UN No.: 3264
MDL No.: MFCD00011286


Certificaten: Traceable to NIST.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR