Barium hydroxide octahydrate 98+% ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

14499.30 14499.A1
14499.30EA 40.8 EUR
14499.30 14499.A1
Barium hydroxide octahydrate 98+% ACS
Barium hydroxide octahydrate

Purity: 98+% CAS No.: 12230-71-6 Molecular Formula: BaH2O2.8H2O Formula Weight: 315.48 (171.36anhy) Physical Form: Crystalline Density: 2.18 Appearance: White Odour: Odourless Melting Point: 78° Solubility: Soluble in water, methanol. Slightly soluble in ethanol. Practically insoluble in acetone Merck Reference: 14,977 UN No.: 3262 MDL No.: MFCD00149152


Used in manufacture of alkali, glass; in synthetic rubber vulcanization, alkalizing agent in water softening, boiler scale remedy, softening water

Formule: Ba(OH)₂·8H₂O
Kookpunt: 780 °C (1013 hPa)
Smeltpunt: 78 °C
Dichtheid: 2,18
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149152
CAS nummer: 12230-71-6
EINECS: 241-234-5
UN: 3262
ADR: 8,III
Merck Index: 14,977

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR