Barium chromate ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

14669.30 14669.A1
14669.30EA 73 EUR
14669.30 14669.A1
Barium chromate ≥98%
Barium chromate

Purity: 98% CAS No.: 10294-40-3 Molecular Formula: BaCrO4 Formula Weight: 253.33 Physical Form: Powder Density: 4.50 Appearance: Yellow Odour: Odourless Solubility: Insoluble in water, acetic acid, chromic acid Merck Reference: 14,972 UN No.: 1479 MDL No.: MFCD00014180

Formule: BaCrO₄
Smeltpunt: 1400 °C
Dichtheid: 4,50
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00014180
CAS nummer: 10294-40-3
EINECS: 233-660-5
UN: 1479
ADR: 5.1,III
Merck Index: 14,972

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR