Lead(II) zirconate 99,7 Metals basis exHf Hf_1

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

14682.22 14682.36
14682.22EA 204 EUR
14682.22 14682.36
Lead(II) zirconate 99,7 Metals basis exHf Hf_1
Lead(II) zirconate

Purity: 99.7% CAS No.: 12060-01-4 Molecular Formula: PbZrO3 Formula Weight: 346.42 Physical Form: -325 Mesh Powder Density: 7.0 Odour: Odourless UN No.: UN3077 MDL No.: MFCD00016275

Formule: O₃PbZr
MW: 346.421 g/mol
Dichtheid: 7.0
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00016275
CAS nummer: 12060-01-4
EINECS: 235-039-4
UN: 3077
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR