Gold ≥99.999% (metals basis), wire, Premion®, Ø 2.0 mm (0.08 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

14733.BO 14733.BQ 14733.BU
14733.BOEA 174 EUR
14733.BO 14733.BQ 14733.BU
Gold ≥99.999% (metals basis), wire, Premion®, Ø 2.0 mm (0.08 in)
Gold

Purity: 99.999% CAS No.: 7440-57-5 Physical Form: ≈60.6g/m Odour: Odourless MDL No.: MFCD00003436

Formule: Au
MW: 196.97 g/mol
Kookpunt: 2800
Smeltpunt: 1063
Dichtheid: 19.3 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003436
CAS nummer: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR