Acidic cleaner and neutraliser, deconex® 26 PLUS
Leverancier: Borer Chemie
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
502300.00-KK5R 502300.00-K10R 502300.00-K25R
148-5265EA 113 EUR
148-5265 148-5266 DECO502300.00-K25R
Acidic cleaner and neutraliser, deconex® 26 PLUS
Reinigingsmiddelen Detergenten en oppervlaktereinigers
Liquid, highly acidic cleaning concentrate und neutraliser for automated cleaning of laboratory glassware und reusable laboratory material.
  • Rinsable without residues and guarantees consistent and error-free results
  • Neutralises carried over alkaline residues
  • Substantially reduces rinsing time and consumption of water
  • In combination with all alkaline deconex® cleaning products, can be also used as a precleaner to remove acid soluble deposits
  • Removes lime residues in laboratory washers
  • Ideal for an effective cleaning of animal cages, also suitable for use in
More Product Information