Draining racks, Poxygrid®, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
H188050000
149-0197EA 320 EUR
149-0197
Draining racks, Poxygrid®, SP Bel-Art
Rekken Afdruiprekken
Racks for drying test tubes, beakers, flasks, and other labware. The stand has rubber tipped feet for non-skid use on a tabletop, or wire loops on the back for wall mounting.
  • Three rows of epoxy-coated hairpin-loop supports (14 in all) hold labware with 18 mm int.Ø necks or larger
  • Top row has 12 straight pins 102 mm long to accommodate smaller items
  • Polystyrene tray catches water as the labware drains