Brushes, swivel-head, for flasks
Leverancier: KOEHLER TECHNISCHE PRODUKTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
K-2500
149-1210EA 27 EUR
149-1210
Brushes, swivel-head, for flasks
Borstels
For intensive and gentle cleaning. Fits flasks from 500 to 5000 ml with SGJ/NS 29 openings.