Gas detection tubes for water vapour

Leverancier: Dräger
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

8101321 8101781 CH23401 8103061
151-0170EA 132 EUR
151-0170 151-0180 151-3152 DRAE8103061
Gas detection tubes for water vapour
Gasbuisjes
To measure the 'spot concentrations' of water vapour for use with an Accuro® pump.

Uses include determination and measurement of concentration peaks, measurement of personal exposure in the inhalation area, detection of leaks in pipelines or gas installations.

One stroke is equivalent to a volume of 100 ±5 ml of gas passing through the tube.

The standard measuring range is calibrated at 20 °C and 1013 hPa.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR