Gas detection tubes for carbon dioxide
Leverancier: Dräger
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
CH23501 CH31401 CH25101 CH20301 8101811
151-0193EA 91.7 EUR
151-0193 151-3147 151-0197 151-0189 151-0181
Gas detection tubes for carbon dioxide
Gasbuisjes
To measure the 'spot concentrations' of carbon dioxide for use with an Accuro® pump. Uses include determination and measurement of concentration peaks, personal exposure in the inhalation area, detection of leaks in pipelines or gas installations, air analysis in sewers, shafts and confined spaces.
One stroke is equivalent to a volume of 100 ±5 ml of gas passing through the tube.