N,O-Bis(trimethylsilyl)acetamide ≥98.5% (by GC), LiChropur™ for GC derivatisation, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

15269-25ML 15269-5ML 15269-10X1ML
15269-25MLEA 212 EUR
15269-25ML 15269-5ML 15269-10X1ML
N,O-Bis(trimethylsilyl)acetamide ≥98.5% (by GC), LiChropur™ for GC derivatisation, Supelco®
N,O-Bis(trimethylsilyl)acetamide
Formule: C₈H₂₁NOSi₂
Smeltpunt: ∼24 °C
MDL Nummer: MFCD00008270
CAS nummer: 10416-59-8
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0a40f8f42759b8bc90a1c4436b6b05137a0607e&vid=b0a40f8f42759b8bc90a1c4436b6b05137a0607e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR