Silicon Standard for ICP, TraceCERT®, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
15747-100ML 08729-100ML
15747-100MLEA 103 EUR
15747-100ML 08729-100ML
Silicon Standard for ICP, TraceCERT®, Supelco®
Standaarden Kationische standaarden Silicium standaarden
Certificaten: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034
Order Now
Specificaties
Environmentally Preferable
M-Clarity quality level MQ300