n-Pentaan 98%, reagent

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Pentane

158941-2.5L 158941-1L
158941-2.5LEA 151 EUR
158941-2.5L 158941-1L
n-Pentaan 98%, reagent
n-Pentaan

  • ≥95% for general laboratory use.


Beilstein Number: 969132
RTECS Number: RZ9450000
UNSPSC Code: 12191502

Formule: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Kookpunt: 36…37 °C
Smeltpunt: –131 °C
Dichtheid: 0.631 g/cm³ (15 °C)
Vlampunt: <–35 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009498
CAS nummer: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3 II
Bereik: 01-2119459286-30
Merck Index: 13,07193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Vapor Density 2.48 (vs air)
Assay (GC) Min. 97.5 %
Refractive index (n 20/D) 1.356 - 1.360
IR spectrum complying
Refractive Index n20/D 1.358 (lit.)
Explosion Limit ~8.3 %
Autoignition Temperature 500 °F
Vapor Pressure 8.43 psi ( 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR