Tetrabutylammonium tribromide ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Tetra-n-butylammonium tribromide, N,N,N,N-Tetrabutylammonium tribromide

16258.14 16258.22
16258.14EA 78.8 EUR
16258.14 16258.22
Tetrabutylammonium tribromide ≥98%
Tetrabutylammonium tribromide

Purity: 98% CAS No.: 38932-80-8 Molecular Formula: C16H36Br3N Formula Weight: 482.18 Physical Form: Crystalline Appearance: Yellow Melting Point: 74-76° Beilstein Reference: 3746114 UN No.: 3261 MDL No.: MFCD00012110

Formule: C₁₆H₃₆Br₃N
MW: 482.18 g/mol
Smeltpunt: 74…76 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012110
CAS nummer: 38932-80-8
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR