Lanthanum(III) carbonate hydrate ≥99.9% (REO, rare earth oxide basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

16639.14 16639.22 16639.36
16639.14EA 22.9 EUR
16639.14 16639.22 16639.36
Lanthanum(III) carbonate hydrate ≥99.9% (REO, rare earth oxide basis)
Lanthanum(III) carbonate hydrate

Purity: 99.9% CAS No.: 54451-24-0 Molecular Formula: La2(CO3)3.xH2O Formula Weight: 457.85(anhy) Physical Form: Powder Density: 2.6-2.7 Merck Reference: 14,5364 MDL No.: MFCD00011069

Formule: La₂(CO₃)₃.xH₂O
Dichtheid: 2,6…2.7
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011069
CAS nummer: 54451-24-0
EINECS: 209-599-5
Merck Index: 14,5364

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR