Hydrazine hydrate 99+%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Hydrazinium hydroxide, Hydrazine monohydrate

16651.22 16651.36 16651.A3 16651.57
16651.22EA 50.5 EUR
16651.22 16651.36 16651.A3 16651.57
Hydrazine hydrate 99+%
Hydrazine hydrate

Hydrazine Hydrate used as a reactant in the cyclizations of pyridinones. It is also used in the study of nanocrystal semiconductors, participating in the functionalization and passivation of surface states.


  • 64% H₂NNH₂ in H₂O


It is widely used as a reducing agent or an intermediate of synthesis in various industrial sectors like water treatment (effluents, industrial boilers), chemical treatment process (metals, mine extraction) or active ingredients synthesis (pharmaceuticals and agrochemicals).

Formule: NH₂NH₂·xH₂O
MW: 32.05 g/mol (anhy)
Kookpunt: 120…121 °C
Smeltpunt: –52 °C
Dichtheid: 1.027
Vlampunt: 75 °C (167 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149931
CAS nummer: 10217-52-4
EINECS: 206-114-9
UN: 2030
ADR: 8 II
Merck Index: 14,4771

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR