Potassium-tert-butoxide 98+%, pure

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

168880050 168881000 168885000 168880250 168880025
168880050.EA 33.1 EUR
168880050. 168881000. 168885000. 168880250. 168880025.
Potassium-tert-butoxide 98+%, pure
Potassium-tert-butoxide
Formule: C₄H₉KO
Smeltpunt: 240…258 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012162
CAS nummer: 865-47-4
EINECS: 212-740-3
UN: 3206
ADR: 4.2,II

Order Now

Specification Test Results

Appearance White to off-white Powder
Titration with HCl ≥98.0 %
KOH + K2CO3 (as KOH) ≤0.50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR