Superose™ 6 Gel Filtration Media, Prep Grade
Leverancier: Cytiva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
17-0489-01
17-0489-01EA 768 EUR
17-0489-01
Superose™ 6 Gel Filtration Media, Prep Grade
Eiwitzuivering Protein Purification Resins
Superose™ 6 PG for separations of proteins and other biomolecules according to size. Wide separation in two molecular weight ranges from 1000 up to 5 million.
  • Achieve high resolution separations across exceptionally broad molecular weight ranges
  • Excellent reproducibility and durability
  • High performance gel filtration media specially designed for preparative purification of biomolecules
Low ionic strength eluents (<0,05 M) can utilise a hydrophobic interaction component to alter the selectivity of Superose™ for some lipids, peptides and small aromatic compounds.