Affinity purification media, GammaBind™ Plus Sepharose™

Leverancier: Cytiva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

17-0886-01 17-0886-02 17-0886-04
17-0886-01EA 1060 EUR
17-0886-01 17-0886-02 17-0886-04
Affinity purification media, GammaBind™ Plus Sepharose™
Eiwitzuivering Protein Purification Resins

GammaBind™ Plus Sepharose™ is GammaBind G, type 3, covalently immobilised to Sepharose™ CL-6B by malimide linkage.


  • For affinity chromatography
  • Recombinant Protein G type 3 coupled to Sepharose CL-6B for enhanced binding to rat and mouse IgG subclasses at laboratory scale
  • Binds to all IgG subclasses from human, mouse, and rat; binds total IgG from guinea pig, rabbit, goat, cow, sheep, and horse
  • Maleimide activation coupling method
  • Yields up to 18 mg/mL mouse IgG
  • Stable in all commonly used aqueous buffers


GammaBind G, Type 3, a recombinant form of streptococcal protein G, binds to the Fc region of IgG from a variety of mammalian species. GammaBind Plus Sepharose may be used to analyze and purify classes, subclasses and fragments of immunoglobulins from any biological fluid or cell culture medium. Since only the Fc region is involved in binding, the Fab region is still available for binding antigen.

Order Now

Specificaties

Ligand Recombinant GammaBind G type 3 lacking albumin-binding region
Environmentally Preferable
Matrix Cross-linked agarose, 6%
Debiet <130 cm/h
pH-bereik 3 - 9
Bindingscapaciteit > 35 mg human IgG/mL
Opslagtemperatuur 2 - 8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR