Ultrapure water systems, Barnstead™ Smart2Pure™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
50129688 50129845 50129885 50129887 50129890 50157872 50157873 50157877 50157882
171-1033EA 7900 EUR
171-1033 171-1034 171-1028 171-1035 171-1029 171-1295 171-0993 171-0994 171-0995
Ultrapure water systems, Barnstead™ Smart2Pure™
Waterzuiveringssystemen Puur- en ultrapuur-watersystemen
The Barnstead Smart2Pure™ system is a compact system that converts tap water into both ASTM Type I and Type II water. It produces 3 or 6 liters of ultra-pure water per hour.
  • Validated 0,2 µm final filter can be autoclaved up to 5 times
  • UV-Oxidation reduces organic compounds in the water to ultra-low levels as well as microorganisms and their metabolites (on UV systems only)
  • Internal ultrafilter (UF) is flushed automatically and has a large surface area to ensure the highest retention of endotoxins and nucleases (on UF systems only)
  • Dispensing is easy and
More Product Information